![Tetrakis[tris(dimethylamino)phosphoranylidenamino]phosphonium chloride 95%](/deepweb/assets/sigmaaldrich/product/structures/160/963/9dd6d457-17b2-44dc-8ea2-d3c0475b3664/640/9dd6d457-17b2-44dc-8ea2-d3c0475b3664.png)
Tetrakis[tris(dimethylamino)phosphoranylidenamino]phosphonium chloride
![Tetrakis[tris(dimethylamino)phosphoranylidenamino]phosphonium chloride 95%](/deepweb/assets/sigmaaldrich/product/structures/160/963/9dd6d457-17b2-44dc-8ea2-d3c0475b3664/640/9dd6d457-17b2-44dc-8ea2-d3c0475b3664.png)
Synonym(s):
Phosphazenium chloride
Linear Formula:
[[(CH3)2N]3P=N]4PCl
CAS Number:
Molecular Weight:
775.26
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing