![Streptolysin O from Streptococcus pyogenes γ-irradiated](/deepweb/assets/sigmaaldrich/product/images/723/601/c203e531-d478-49fe-9304-3aac2b9e499a/640/c203e531-d478-49fe-9304-3aac2b9e499a.jpg)
Streptolysin O from Streptococcus pyogenes
![Streptolysin O from Streptococcus pyogenes γ-irradiated](/deepweb/assets/sigmaaldrich/product/images/723/601/c203e531-d478-49fe-9304-3aac2b9e499a/640/c203e531-d478-49fe-9304-3aac2b9e499a.jpg)
Synonym(s):
Streptolysin O from Streptococcus pyogenes
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Purity
Activity Spectrum
Research Area
Chemical Composition
Quality Segment
Application
Mode of Action
Sterilization Method
Showing 1-3 of 3
Compare
Product Number
Description
Pricing