Skip to Content
Merck
Sodium nitroprusside dihydrate [disodium pentacyanonitrosyl ferrate(III) dihydrate] GR for analysis ACS,Reag. Ph Eur

Sodium nitroprusside dihydrate

Sodium nitroprusside dihydrate [disodium pentacyanonitrosyl ferrate(III) dihydrate] GR for analysis ACS,Reag. Ph Eur
Synonym(s):
Sodium nitroprusside dihydrate
Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
281.95

Products

Brand
Formula Weight
Agency
Analyte Suitability
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing