Skip to Content
Merck
Sodium ionophore III Selectophore™, function tested

Sodium ionophore III

Sodium ionophore III Selectophore™, function tested
Synonym(s):
ETH 2120, N,N,N′,N′-Tetracyclohexyl-1,2-phenylenedioxydiacetamide
Empirical Formula (Hill Notation):
C34H52N2O4
CAS Number:
Molecular Weight:
552.79

Products

Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing