
Sodium ferrocyanide decahydrate

Synonym(s):
Sodium hexacyanoferrate(II)
Linear Formula:
Na4Fe(CN)6 · 10H2O
CAS Number:
Molecular Weight:
484.06
Products
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing