![Saquinavir mesilate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/393/316/77953fb1-1a02-4566-b558-4c2c9fd282ea/640/77953fb1-1a02-4566-b558-4c2c9fd282ea.png)
Saquinavir mesilate
![Saquinavir mesilate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/393/316/77953fb1-1a02-4566-b558-4c2c9fd282ea/640/77953fb1-1a02-4566-b558-4c2c9fd282ea.png)
Synonym(s):
Saquinavir mesylate, (2S)-N1[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-Dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]butanediamide methanesulfonate, Ro-31-8959
Empirical Formula (Hill Notation):
C38H50N6O5·CH4O3S
CAS Number:
Molecular Weight:
766.95
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing