Salicylic acid (phenyl-13C6)
Synonym(s):
2-Hydroxybenzoic acid (phenyl-13C6)
Linear Formula:
2-(HO)13C6H4CO2H
CAS Number:
Molecular Weight:
144.08
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing