(RS)-Mevalonic acid lithium salt
Synonym(s):
(±)-MVA-Li, rac.-MVA-Li, Lithium (±)-3,5-dihydroxy-3-methyl-pentanoate, Lithium (±)-3,5-dihydroxy-3-methyl-valerate
Empirical Formula (Hill Notation):
C6H11O4 · xLi+
CAS Number:
Molecular Weight:
147.15 (free acid basis)
Products
Brand
Formula Weight
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing