
R-Omeprazole

Synonym(s):
5-Methoxy-2-[(R-[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]sulfinyl]-1H-benzimidazole sodium salt
Empirical Formula (Hill Notation):
C17H18N3NaO3S
CAS Number:
Molecular Weight:
367.40
Products
Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing