Resorufin methyl ether
Synonym(s):
7-Methoxy-3H-phenoxazin-3-one, Methoxyresorufin, O7-Methylresorufin
Empirical Formula (Hill Notation):
C13H9NO3
CAS Number:
Molecular Weight:
227.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing