
Rescinnamine

Synonym(s):
Methyl reserpate 3,4,5-trimethoxycinnamic acid ester, Reserpinine
Empirical Formula (Hill Notation):
C35H42N2O9
CAS Number:
Molecular Weight:
634.72
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing