Skip to Content
Merck
rAlbumin human United States Pharmacopeia (USP) Reference Standard

rAlbumin human

rAlbumin human United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Albumin human, rHSA
Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53

Products

Product Category
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing