Skip to Content
Merck
(R)-1-Boc-3-pyrrolidinecarboxylic acid 97%

(R)-1-Boc-3-pyrrolidinecarboxylic acid

(R)-1-Boc-3-pyrrolidinecarboxylic acid 97%
Synonym(s):
(R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid, N-Boc-L-β-proline
Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing