Pyrimethanil
Synonym(s):
2-Anilino-4,6-dimethylpyrimidine
Empirical Formula (Hill Notation):
C12H13N3
CAS Number:
Molecular Weight:
199.25
Products
Product Category
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2