Skip to Content
Merck
Pyridoxal phosphate-6-azo(benzene-2,4-disulfonic acid) tetrasodium salt hydrate solid, ≥98% (HPLC)

Pyridoxal phosphate-6-azo(benzene-2,4-disulfonic acid) tetrasodium salt hydrate

Pyridoxal phosphate-6-azo(benzene-2,4-disulfonic acid) tetrasodium salt hydrate solid, ≥98% (HPLC)
Synonym(s):
4-[[4-Formyl-5-hydroxy-6-methyl-3-[(phosphonooxy)methyl]-2-pyridinyl]azo]-1,3-benzenedisulfonic acid tetrasodium salt, PPADS tetrasodium salt hydrate
Empirical Formula (Hill Notation):
C14H10N3Na4O12PS2 · xH2O
CAS Number:
Molecular Weight:
599.31 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1