Skip to Content
Merck
Pyridine-2-aldehyde-DNPH solution 287 μg/mL in acetonitrile: DMSO (7:3), certified reference material, ampule of 2 mL, Cerilliant®

Pyridine-2-aldehyde-DNPH solution

Pyridine-2-aldehyde-DNPH solution 287 μg/mL in acetonitrile: DMSO (7:3), certified reference material, ampule of 2 mL, Cerilliant®
Synonym(s):
Pyridine-2-aldehyde-DNPH solution
Empirical Formula (Hill Notation):
C12H9N5O4
CAS Number:
Molecular Weight:
287.23

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade