![Plastic additive 12 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/666/56f07312-355e-4612-b63c-304d69d2c06b/640/56f07312-355e-4612-b63c-304d69d2c06b.png)
Plastic additive 12
![Plastic additive 12 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/666/56f07312-355e-4612-b63c-304d69d2c06b/640/56f07312-355e-4612-b63c-304d69d2c06b.png)
Synonym(s):
Tris(2,4-di-tert-butylphenyl) phosphite
Linear Formula:
[[(CH3)3C]2C6H3O]3P
CAS Number:
Molecular Weight:
646.92
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing