Plastic additive 01
Synonym(s):
Bis(2-ethylhexyl) phthalate, DEHP, Phthalic acid bis(2-ethylhexyl ester), ‘Dioctyl’ phthalate
Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing