Pirimicarb
Synonym(s):
2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate
Empirical Formula (Hill Notation):
C11H18N4O2
CAS Number:
Molecular Weight:
238.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2