
Phosphomycin disodium salt

Synonym(s):
(−)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, Fosfomycin, Phosphonomycin
Empirical Formula (Hill Notation):
C3H5Na2O4P
Molecular Weight:
182.02
EC Number:
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing