Skip to Content
Merck
Phosphocreatine disodium salt hydrate ≥97%

Phosphocreatine disodium salt hydrate

Phosphocreatine disodium salt hydrate ≥97%
Synonym(s):
N-(Imino[phosphonoamino]methyl)-N-methylglycine
Linear Formula:
C4H8N3O5PNa2 · x H2O
CAS Number:
Molecular Weight:
255.08 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing