![Phenylephrine Bitartrate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/326/215/95772b42-c6f1-459c-99ef-72ae9c56eb44/640/95772b42-c6f1-459c-99ef-72ae9c56eb44.jpg)
Phenylephrine bitartrate
![Phenylephrine Bitartrate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/326/215/95772b42-c6f1-459c-99ef-72ae9c56eb44/640/95772b42-c6f1-459c-99ef-72ae9c56eb44.jpg)
Synonym(s):
(R-3-Hydroxy-α-(methylaminomethyl)benzyL alcohol hydrogen L-tartrate
Empirical Formula (Hill Notation):
C9H13NO2 · C4H6O6
CAS Number:
Molecular Weight:
317.29
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2