Skip to Content
Merck
Palmitic acid N-hydroxysuccinimide ester ≥98% (TLC)

Palmitic acid N-hydroxysuccinimide ester

Palmitic acid N-hydroxysuccinimide ester ≥98% (TLC)
Synonym(s):
N-(Palmitoyloxy)succinimide, N-Succinimidyl palmitate, Palmitic acid N-succinimidyl ester
Empirical Formula (Hill Notation):
C20H35NO4
CAS Number:
Molecular Weight:
353.50

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing