
Paclitaxel impurity Q

Synonym(s):
N-Debenzoyl-N-[(3E)-hex-3-enoyl]paclitaxel
Empirical Formula (Hill Notation):
C46H55NO14
CAS Number:
Molecular Weight:
845.93
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing