Skip to Content
Merck
o-Terphenyl 99%

o-Terphenyl

o-Terphenyl 99%
Synonym(s):
1,2-Diphenylbenzene, o-Terphenyl
Linear Formula:
2-(C6H5)C6H4C6H5
CAS Number:
Molecular Weight:
230.30

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing