Skip to Content
Merck
vibrant-m

Osmium(VIII) oxide

vibrant-m
Synonym(s):
Osmium(VIII) oxide
Empirical Formula (Hill Notation):
O4Os
CAS Number:
Molecular Weight:
254.23

Products

Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
pH Value
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing