![Norelgestromin certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/342/779/f7a13ea2-9737-4c38-8d85-8e9e199e2ff4/640/f7a13ea2-9737-4c38-8d85-8e9e199e2ff4.jpg)
Norelgestromin
![Norelgestromin certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/342/779/f7a13ea2-9737-4c38-8d85-8e9e199e2ff4/640/f7a13ea2-9737-4c38-8d85-8e9e199e2ff4.jpg)
Synonym(s):
Deacetylnorgestimate, (17alpha)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one oxime
Empirical Formula (Hill Notation):
C21H29NO2
CAS Number:
Molecular Weight:
327.46
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2