
New fuchsin (C.I. 42520)

Empirical Formula (Hill Notation):
C22H24ClN3
CAS Number:
Molecular Weight:
365.90
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing