![Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse clone 24516, purified immunoglobulin, lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/134/894/b8522e60-f68f-4483-9f06-dc8a40632f8e/640/b8522e60-f68f-4483-9f06-dc8a40632f8e.jpg)
Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse
![Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse clone 24516, purified immunoglobulin, lyophilized powder](/deepweb/assets/sigmaaldrich/product/images/134/894/b8522e60-f68f-4483-9f06-dc8a40632f8e/640/b8522e60-f68f-4483-9f06-dc8a40632f8e.jpg)
Synonym(s):
Anti-HGF
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing