
Modafinil for system suitability

Synonym(s):
Modafinil, 2-[(Diphenylmethyl)sulfinyl]acetamide
Empirical Formula (Hill Notation):
C15H15NO2S
CAS Number:
Molecular Weight:
273.35
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing