Skip to Content
Merck
vibrant-m

Methylthioninium for system suitability

vibrant-m
Synonym(s):
Methylthioninium chloride, Methylene blue
Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1