Skip to Content
Merck
Methyl laurate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Methyl laurate

Methyl laurate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Lauric acid methyl ester, Methyl dodecanoate, Methyl laurate
Linear Formula:
CH3(CH2)10CO2CH3
CAS Number:
Molecular Weight:
214.34
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique