![Mesaconitine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/318/569/50d98cbd-e7af-4d93-887e-b4bbc0ee8293/640/50d98cbd-e7af-4d93-887e-b4bbc0ee8293.png)
Mesaconitine
![Mesaconitine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/318/569/50d98cbd-e7af-4d93-887e-b4bbc0ee8293/640/50d98cbd-e7af-4d93-887e-b4bbc0ee8293.png)
Synonym(s):
Mesaconitine, (1a,3a,6a,14a,15a,16β)-1,6,16-Trimethoxy-4-(methoxymethyl)-20-methyl-aconitane-3,8,13,14,15-pentol 8-acetate 14-benzoate
Empirical Formula (Hill Notation):
C33H45NO11
CAS Number:
Molecular Weight:
631.71
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing