
Mechlorethamine hydrochloride

Synonym(s):
2-Chloro-N-(2-chloroethyl)-N-methylethanamine, N-Methylbis(2-chloroethyl)amine hydrochloride, Bis(2-Chloroethyl)methylamine, Bis(2-chloroethyl)methylamine, MBA hydrochloride, NM, Nitrogen mustard
Linear Formula:
(ClCH2CH2)2NCH3·HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1