![Malathion impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/119/823/3f2ca13b-c32d-415c-a848-e477061d5b60/640/3f2ca13b-c32d-415c-a848-e477061d5b60.png)
Malathion impurity B
![Malathion impurity B European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/119/823/3f2ca13b-c32d-415c-a848-e477061d5b60/640/3f2ca13b-c32d-415c-a848-e477061d5b60.png)
Synonym(s):
Malaoxon, Diethyl 2-[(dimethoxyphosphoryl)sulfanyl]succinate
Empirical Formula (Hill Notation):
C10H19O7PS
CAS Number:
Molecular Weight:
314.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing