Skip to Content
Merck
Lypressin United States Pharmacopeia (USP) Reference Standard

Lypressin

Lypressin United States Pharmacopeia (USP) Reference Standard
Synonym(s):
[Lys8]-Vasopressin, Lysine vasopressin, Lyspressin
Empirical Formula (Hill Notation):
C46H65N13O12S2
CAS Number:
Molecular Weight:
1056.22

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing