Skip to Content
Merck
vibrant-m

Lynestrenol for peak identification

vibrant-m
Synonym(s):
Lynestrenol, (17α)-19-Norpregn-4-en-20-yn-17-ol
Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1