Skip to Content
Merck
Lurasidone hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Lurasidone hydrochloride solution

Lurasidone hydrochloride solution 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C28H36N4O2S · HCl
CAS Number:
Molecular Weight:
529.14
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique