Skip to Content
Merck
L-Dihydrocarvyl acetate mixture of isomers, ≥95%

L-Dihydrocarvyl acetate

L-Dihydrocarvyl acetate mixture of isomers, ≥95%
Synonym(s):
(−)-Dihydrocarvyl acetate
Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing