![L-Arginine p-nitroanilide dihydrochloride protease substrate](/deepweb/assets/sigmaaldrich/product/structures/212/479/5622d11a-394c-4c29-8d54-a7dd2a550303/640/5622d11a-394c-4c29-8d54-a7dd2a550303.png)
L-Arginine p-nitroanilide dihydrochloride
![L-Arginine p-nitroanilide dihydrochloride protease substrate](/deepweb/assets/sigmaaldrich/product/structures/212/479/5622d11a-394c-4c29-8d54-a7dd2a550303/640/5622d11a-394c-4c29-8d54-a7dd2a550303.png)
Empirical Formula (Hill Notation):
C12H18N6O3 · 2HCl
CAS Number:
Molecular Weight:
367.23
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing