![Lamotrigine Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/809/663/46e5552c-1776-4e73-af71-8570820afb00/640/46e5552c-1776-4e73-af71-8570820afb00.png)
Lamotrigine Related Compound D
![Lamotrigine Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/809/663/46e5552c-1776-4e73-af71-8570820afb00/640/46e5552c-1776-4e73-af71-8570820afb00.png)
Synonym(s):
N-[5-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide
Empirical Formula (Hill Notation):
C16H9Cl4N5O
CAS Number:
Molecular Weight:
429.09
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2