Isosteviol
Synonym(s):
(−)-Isosteviol, (4α,8β,13β)-13--Methyl-16-oxo-17-norkauran-18-oic acid, Ketoisostevic acid, NSC 231875
Empirical Formula (Hill Notation):
C20H30O3
CAS Number:
Molecular Weight:
318.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing