Skip to Content
Merck
vibrant-m

Isosorbide 2-nitrate

vibrant-m
Empirical Formula (Hill Notation):
C6H9NO6
CAS Number:
Molecular Weight:
191.14

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade