Isopentyl nitrite
Synonym(s):
Isoamyl nitrite, Isopentyl nitrite
Linear Formula:
(CH3)2CHCH2CH2ONO
CAS Number:
Molecular Weight:
117.15
Products
Product Category
Brand
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing