Isoeugenol, predominantly trans
Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing