(Ir[dF(CF3)ppy]2(dtbpy))PF6
Synonym(s):
[Ir{dF(CF3)ppy}2(dtbpy)]PF6
Empirical Formula (Hill Notation):
C42H34F16IrN4P
CAS Number:
Molecular Weight:
1121.91
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing