Skip to Content
Merck
vibrant-m

Iodixanol impurity E

vibrant-m
Synonym(s):
5-[Acetyl[3-[acetyl[3-carbamoyl-5-[(2,3-dihydroxypropyl)carbamoyl]-2,4,6-triiodophenyl]amino]-2-hydroxypropyl]amino]-N,N′-bis(2,3-dihydroxypropyl)-2,4,6-triiodobenzene-1,3-dicarboxamide, Iodixanol amide
Empirical Formula (Hill Notation):
C32H38I6N6O13
CAS Number:
Molecular Weight:
1476.10

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing