Skip to Content
Merck
Indapamide British Pharmacopoeia (BP) Reference Standard

Indapamide

Indapamide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Indapamide, N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline
Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique