![Hyaluronidase from Streptomyces hyalurolyticus](/deepweb/assets/sigmaaldrich/product/images/335/296/cc0da2f4-dd61-4eb3-9c99-c179e0e487cc/640/cc0da2f4-dd61-4eb3-9c99-c179e0e487cc.jpg)
Hyaluronidase from Streptomyces hyalurolyticus
![Hyaluronidase from Streptomyces hyalurolyticus](/deepweb/assets/sigmaaldrich/product/images/335/296/cc0da2f4-dd61-4eb3-9c99-c179e0e487cc/640/cc0da2f4-dd61-4eb3-9c99-c179e0e487cc.jpg)
Synonym(s):
Hyaluronate Lyase from Streptomyces hyalurolyticus
CAS Number:
EC Number:
Products
Brand
Biological Source
Physical Form
Quality Segment
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing