Skip to Content
Merck
Heptaldehyde analytical standard

Heptaldehyde

Heptaldehyde analytical standard
Synonym(s):
1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde
Linear Formula:
CH3(CH2)5CHO
CAS Number:
Molecular Weight:
114.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing